Learn More
Cerivastatin Sodium Salt, TRC
Product Code. 30133010
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133010 25 mg 25mg
30135341 5 mg 5mg
30003341 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133010 Supplier Toronto Research Chemicals Supplier No. TRCC27700025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 143201-11-0
Molecular Formula C26 H33 F N O5 . Na
Molecular Weight (g/mol) 481.53
Synonym 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-5-(methoxymethyl)-2,6-bis(1-methylethyl)-3-pyridinyl]-3,5-dihydroxy-, sodium salt (1:1), (3R,5S,6E)-, 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-5-(methoxymethyl)-2,6-bis(1-methylethyl)-3-pyridinyl]-3,5-dihydroxy-, monosodium salt, (3R,5S,6E)- (9CI), 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-5-(methoxymethyl)-2,6-bis(1-methylethyl)-3-pyridinyl]-3,5-dihydroxy-, monosodium salt, [S-[R*,S*-(E)]]-, BAY-w 6228, Baycol, Cerivastatin sodium, Lipobay, Rivastatin
IUPAC Name sodium;(E,3R,5S)-7-[4-(4-fluorophenyl)-5-(methoxymethyl)-2,6-di(propan-2-yl)pyridin-3-yl]-3,5-dihydroxyhept-6-enoate
SMILES [Na+].COCc1c(nc(C(C)C)c(\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])c1c2ccc(F)cc2)C(C)C
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C26H34FNO5.Na/c1-15(2)25-21(11-10-19(29)12-20(30)13-23(31)32)24(17-6-8-18(27)9-7-17)22(14-33-5)26(28-25)16(3)4;/h6-11,15-16,19-20,29-30H,12-14H2,1-5H3,(H,31,32);/q;+1/p-1/b11-10+;/t19-,20-;/m1./s1
Formula Weight 481.224
Percent Purity >95%
Chemical Name or Material Cerivastatin, Sodium Salt
Grade HPLC