missing translation for 'onlineSavingsMsg'
Learn More

Cepharanthine, TRC

Product Code. 30367882
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367882 500 mg 500mg
30362737 100 mg 100mg
30366962 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30367882 Supplier Toronto Research Chemicals Supplier No. TRCC261513500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 481-49-2
Molecular Formula C37 H38 N2 O6
Molecular Weight (g/mol) 606.71
Synonym 1H-4,6:16,19-Dietheno-21,25-metheno-12H-[1,3]dioxolo[4,5-g]pyrido[2',3':17,18][1,10]dioxacycloeicosino[2,3,4-ij]isoquinoline, 2,3,13,14,14a,15,26,26a-octahydro-22,30-dimethoxy-1,14-dimethyl-, (14aS,26aR)-, Cepharanthine (6CI,7CI,8CI), Oxyacanthan, 6',12'-dimethoxy-2,2'-dimethyl-6,7-[methylenebis(oxy)]-, (+)-Cepharanthine, 1H-4,6:16,19-Dietheno-21,25-metheno-12H-[1,3]dioxolo[4,5-g]pyrido[2',3':17,18][1,10]dioxacycloeicosino[2,3,4-ij]isoquinoline, 2,3,13,14,14a,15,26,26a-octahydro-22,30-dimethoxy-1,14-dimethyl-, [14aS-(14aR*,26aS*)]-, Cepharanthin, Cepharantin, NSC 623442, O-Methylcepharanoline
IUPAC Name (14S,27R)-22,33-dimethoxy-13,28-dimethyl-2,5,7,20-tetraoxa-13,28-diazaoctacyclo[25.6.2.216,19.13,10.121,25.04,8.031,35.014,39]nonatriaconta-1(33),3(39),4(8),9,16(38),17,19(37),21,23,25(36),31,34-dodecaene
SMILES COc1ccc2C[C@H]3N(C)CCc4cc(OC)c(Oc5c6OCOc6cc7CCN(C)[C@@H](Cc8ccc(Oc1c2)cc8)c57)cc34

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C37H38N2O6/c1-38-13-11-24-18-31(41-4)33-20-27(24)28(38)16-23-7-10-30(40-3)32(17-23)44-26-8-5-22(6-9-26)15-29-35-25(12-14-39(29)2)19-34-36(37(35)45-33)43-21-42-34/h5-10,17-20,28-29H,11-16,21H2,1-4H3/t28-,29+/m1/s1
Formula Weight 606.273
Chemical Name or Material Cepharanthine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.