Learn More
Eppendorf™ Centrifuge Adapters For Rotors for 5810 and 5810R
Available in a variety of configurations to suit a wide range of applications
179.00 EUR - 208.00 EUR
Specifications
Type | Adapter |
---|---|
For Use With (Equipment) | Rotors 5810 and 5810R Centrifuge |
No. per Pack | 2 Pack |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
10293172
|
Eppendorf™
5810750008 |
1.2 to 5 mL |
179.00 EUR
Pack of 2 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11518623
|
Eppendorf™
5810751004 |
1.5 to 2 mL |
179.00 EUR
Pack of 2 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10554332
|
Eppendorf™
5810755000 |
15 mL |
203.00 EUR
Pack of 2 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10326802
|
Eppendorf™
5810770009 |
180 to 250 mL |
208.00 EUR
Pack of 2 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10134961
|
Eppendorf™
5810752000 |
2.6 to 7 mL |
188.00 EUR
Pack of 2 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11528623
|
Eppendorf™
5810753007 |
3 to 15 mL |
188.00 EUR
Pack of 2 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11548623
|
Eppendorf™
5810759005 |
30 to 50 mL |
179.00 EUR
Pack of 2 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10776695
|
Eppendorf™
5810757002 |
18 to 30 mL |
179.00 EUR
Pack of 2 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10102082
|
Eppendorf™
5810760003 |
50 to 75 mL |
179.00 EUR
Pack of 2 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10106771
|
Eppendorf™
5810758009 |
50 mL |
203.00 EUR
Pack of 2 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10253222
|
Eppendorf™
5810756006 |
7 to 18 mL |
179.00 EUR
Pack of 2 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11558623
|
Eppendorf™
5810761000 |
80 to 120 mL |
179.00 EUR
Pack of 2 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Adaptors for Eppendorf™ Centrifuges 5810 and 5810R.
- Compatible with conical or flat tubes
Specifications
Adapter | |
Rotors 5810 and 5810R Centrifuge | |
2 Pack |