missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cenerimod, MedChemExpress
Description
Cenerimod (ACT-334441) is a potent, selective and orally active S1P1 receptor modulator, with an EC50 of 1 nM. Cenerimod shows more than 36‐fold selctivity for hS1P1 over hS1P2, hS1P3, hS1P4, and hS1P5 receptor subtypes (EC50s=>10000, 228, 2134, and 36 nM, respectively). Cenerimod can attenuate murine experimental autoimmune encephalomyelitis (EAE) and murine sclerodermatous.
Specifications
Specifications
| Chemical Name or Material | Cenerimod |
| CAS | 1262414-04-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H31N3O5 |
| Quantity | 10 mg |
| Synonym | ACT-334441 |
| Solubility Information | DMSO : ≥ 100 mg/mL (220.49 mM) |
| SMILES | OC[C@H](O)COC1=C(C)C=C(C2=NOC(C3=CC(C4CCCC4)=NC(OC)=C3)=N2)C=C1CC |
| Molecular Weight (g/mol) | 453.53 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?