Learn More
Eppendorf™ CellXpert™ C170i
Product Code. 15937065
Click to view available options
Voltage:
220/240 V
Unit Size:
Each
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 15937065
Brand: Eppendorf™ 6731000011
Only null left
Add to basket
Add to basket
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
The CellXpert C170i CO2 incubator provides reliable growth conditions (fast recovery, uniform temperature, fanless design) for more reproducible results and the fanless design makes cleaning easy. It also provides up to 25 % more usable space and many smart features with the VisioNize™ touch interface.
  • Non-segmented inner door
  • 180° HTD (standard with C170i)
  • 220 – 240 V, 50 – 60 Hz
TRUSTED_SUSTAINABILITY
Specifications
CO2 Concentration Control ±0.1 %
Controller Type VisioNize™ touch interface
Data Outputs USB, Ethernet, VisioNize™ onboard
Capacity (Metric) 170 L
CO2 Concentration Range 0.1 to 20%
CO2 Sensor Technology IR Sensor
No. of Shelves 3 std./8 max.
Plug Type EU
Temperature Range (Metric) Ambient +4° to 50°C
Type CO2 Incubator
Door Style outer door with magnetic closure
Ports 2
Sample Volume (Metric) 170 L
Temperature Stability Ambient +4 °C to 50 °C
Dimensions (D x W x H) Interior 44.5 x 53.9 x 69.2 cm
Dimensions (L x W x H) 70.5 x 71.8 x 90 cm
Width (Metric) Interior 53.9 cm
Height (Metric) Interior 69.2 cm
Depth (Metric) Interior 44.5 cm
Weight (Metric) 81 kg
Voltage 220/240 V