Learn More
Sartorius™ Cellulose / Polyethylene Coated Surface Protection Paper
Product Code. 11724986 Shop All Sartorius Products
Click to view available options
Length (Metric):
100 m
50 m
Width (Metric):
400 mm
460 mm
600 mm
Unit Size:
Each
Product Code. 11724986
Brand: Sartorius™ FT1601400100
Specifications
Product Type Surface Protection Paper
Material Cellulose/Polyethylene
Absorbancy 125 mL/m2
Length (Metric) 100 m
Width (Metric) 400 mm
Color White