Learn More
Ceftiofur Hydrochloride, TRC
Product Code. 17911305
Change view
Click to view available options
Quantity:
50 mg
500 mg
5 g
Unit Size:
500mg
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17911305 50 mg 50mg
17941035 5 g 5g
17934605 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17911305 Supplier Toronto Research Chemicals Supplier No. TRCC24470050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103980-44-5
Molecular Formula C19 H17 N5 O7 S3 . Cl H
Molecular Weight (g/mol) 560.02
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-[[(2-furanylcarbonyl)thio]methyl]-8-oxo-, monohydrochloride, [6R-[6α,7β(Z)]]-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-[[(2-furanylcarbonyl)thio]methyl]-8-oxo-, monohydrochloride, (6R,7R)- (9CI), Aviar, Cefalus, Ceftiofur hydrochloride, Excenel, Excenel RTU, Spectramast LC, U 64279A, U 67279A
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(furan-2-carbonylsulfanylmethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrochloride
SMILES Cl.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)CSC(=O)c3occc3)\c4csc(N)n4
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C19H17N5O7S3.ClH/c1-30-23-11(9-7-34-19(20)21-9)14(25)22-12-15(26)24-13(17(27)28)8(5-32-16(12)24)6-33-18(29)10-3-2-4-31-10;/h2-4,7,12,16H,5-6H2,1H3,(H2,20,21)(H,22,25)(H,27,28);1H/b23-11-;/t12-,16-;/m1./s1
Formula Weight 559.0057
Percent Purity >95%
Chemical Name or Material Ceftiofur hydrochloride
Grade HPLC