Learn More
Cefoperazone sodium salt, MP Biomedicals™
Product Code. 11455335 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11455335
Brand: MP Biomedicals 0219969505
Cefoperazone sodium salt USP, White powder
  • It is used to study drug-protein binding, expression and inhibition of penicillin-binding proteins (PBPs) during cell wall synthesis.
  • Cefoperazone sodium salt is a broad spectrum third generation cephalosporin antibiotic that has similar antimicrobial activity to ceftazidime.
  • It is active against Pseudomonoas aeruginosa and it has enhanced activity against Enterobacteriaceae and Bacteroides sp. in the presence of the beta-lactamase inhibitor sulbactam. Compared to cefotaxime, cefoperazone has greater susceptibility to hydrolysis by some beta-lactamases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62893-20-3
Molecular Formula C25H26N9NaO8S2
Molecular Weight (g/mol) 667.65
MDL Number MFCD07793331
InChI Key NCFTXMQPRQZFMZ-WERGMSTESA-M
Specifications
Color White
pH 4.5 to 6.5 (1 in 4)
Quantity 5 g
Beilstein 4902135
Synonym Cefobid, Cefoperazone sodium, CPZ, T-1551, [6R-[6.alpha.,7.beta.(R*)]]-7- [[[[(4-Ethyl-2,3- dioxo-1-piperazinyl) carbonyl]amino] (4-hydroxyphenyl) acetyl] amino]-3- [[(1-methyl-1H-tetrazol-5-yl) thio] methyl]- 8-oxo-5- thia-1-azabicyclo[4.2.0], oct-2-ene-2- carboxylic acid, 7-[D-(-)-alpha- (4-ethyl-2,3-dioxo- 1-piperazinecarboxamido)- alpha-(4-hydroxyphenyl) acetamido]- 3-[[(1-methyl-1H- tetrazol-5-yl) thio] methyl]-3-cephem-4- carboxylic acid
Solubility Information Soluble in water (50mg/mL - clear, faint yellow solution).
Physical Form Powder
compliance-icons compliance-icons
Product Identifier
  • Cefoperazone Sodium Salt, USP
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Cefoperazone SodiumSalt