Learn More
Molecular Probes™ CDP-Star™ Substrate (0.25 mM Ready-To-Use)
Product Code. 10125425
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
Unit Size:
100mL
50mL
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10125425 50 mL 50mL
10144664 100 mL 100mL
12674325 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10125425 Supplier Molecular Probes™ Supplier No. T2145
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The CDP-Star™ Substrate is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes.
The CDP-Star™ Substrate is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.
This CDP-Star™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.
  • Chemiluminescent substrate provides highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
  • Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.
This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours.
TRUSTED_SUSTAINABILITY
Specifications
Description CDP-Star™ Substrate, 50mL, Liquid Form, Chemiluminescent Alkaline Phosphatase Substrate, 0.25mM Ready-to-use For Protein or Nucleic Acid Blotting on Nitrocellulose membranes, Protein Assays and Analysis, Southern, Northern and Western blotting, Can also be used in Solution-based Assays
Product Type ELISA Substrate
Quantity 50 mL
Detection Method Chemiluminescence
Form Liquid
Format Bottle
For Use With (Application) Protein assays and analysis, southern, northern, western blotting, can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays, protein biology, cell analysis, enzyme reporter assays, ELISA, alkaline phosphatase assays, build your own immunoassay, ready-to-use immunoassay
Product Line CDP-Star, NovaBright
Substrate Type Alkaline Phosphatase
Content And Storage Store the product at 4°C.
Shipping Condition Room Temperature