missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDNF Monoclonal Antibody (D2), Invitrogen™

Product Code. 17674346
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17674346 20 μL 20µL
17664346 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17674346 Supplier Invitrogen™ Supplier No. MA543893

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The conserved dopamine neurotrophic factor (CDNF) is a neurotrophic factor for dopaminergic neurons and highly homologous to the mesencephalic-astrocyte-derived neuro-trophic factor. Somewhat surprisingly, CDCF is expressed at higher levels in tissues such as heart, skeletal muscle and testes than in brain. Similar to the glial cell line-derived neurotrophic factor (GDNF), CDNF can prevent the 6-hydroxylamine (6 OHDA) induced degeneration of dopaminergic neurons in a rat model of Parkinsons disease. Furthermore, CDNF was able to restore the dopaminergic function and prevent the degeneration of dopaminergic neurons in the substantia nigra, suggesting that CDNF might be suitable for the treatment of Parkinsons disease. At least two isoforms of CDNF are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDNF
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Cdnf
Gene Accession No. Q49AH0
Gene Alias 9330140G23; arginine-rich protein mutated in early stage tumors-like 1; arginine-rich, mutated in early stage tumors-like 1; ARMETL1; ARMET-like protein 1; Cdnf; Cerebral dopamine neurotrophic factor; conserved dopamine neurotrophic factor
Gene Symbols Cdnf
Host Species Mouse
Immunogen Pro33-Leu187 (Accession Q49AH0), with N-terminal His and GST Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441549
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.