missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CDK18/cyclin Y, GST Tag Recombinant Protein

Product Code. 15980273
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15980273 10 μg 10µg
17188725 100 μg 100µg
17108775 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15980273 Supplier Invitrogen™ Supplier No. A30492

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This CDK18/cyclin Y recombinant human protein (CDK18: full length, cyclin Y: 2-end) was expressed in insect cells. CDK18/cyclin Y recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic The gene accession number for CDK18/cyclin Y are AAH11526 and NP_659449.3.

CDK18/cyclin Y serine/threonine kinase or cyclin-dependent kinase 18 belongs to the cdc2/cdkx subfamily of kinases. CDK18/cyclin Y is thought to play a role in terminally differentiated neurons and it contains an N-terminal extension relative to CDC2. CDK18/cyclin Y is expressed in a variety of human cell lines and tissues. Elevation in the protein levels of CDK18/cyclin Y in the temporal cortex of Alzheimer's disease relative to control brains has been observed. It is proposed that CDK18/cyclin Y is a paired helical filaments-associated kinase that modulates tau phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q07002
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 25% glycerol, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 150mM NaCl, 0.25mM DTT and no preservative; pH 7.5
Gene ID (Entrez) 5129
Molecular Weight (g/mol) 80.5-65.5 kDa
Name Human CDK18/cyclin Y, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias PCTK3
Common Name CDK18
Gene Symbol CDK18
KinaseFamily CDKs and CHKs
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence CDK18: Full Length, Cyclin Y: 2-341
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Active, Catalytic
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.