missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD91 Monoclonal Antibody (A2MR-a2), Super Bright™ 600, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 63-0919-41
This item is not returnable.
View return policy
Description
Mouse Monoclonal Antibody
CD91 Monoclonal antibody specifically detects CD91 in Human samples. It is validated for Flow Cytometry
Specifications
| CD91 | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Q07954 | |
| LRP1 | |
| Affinity chromatography | |
| RUO | |
| 4035 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| A2MR-a2 | |
| Super Bright 600 | |
| LRP1 | |
| A2MR; AI316852; Alpha-2-macroglobulin receptor; APOER; Apolipoprotein E receptor; APR; b2b1554Clo; CD91; FLJ16451; IGFBP3R; Lrp; Lrp1; LRP-1; LRP1A; MGC88725; Prolow-density lipoprotein receptor-related protein 1; TGFBR5 | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction