missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD85h (ILT1) Monoclonal Antibody (135.4), PE, eBioscience™
GREENER_CHOICE

Product Code. 15546996
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15546996 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15546996 Supplier Invitrogen™ Supplier No. 12511942

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Description: The monoclonal antibody 135.4 recognizes ILT1, also known as CD85h, LIR7 and LILRA2. ILT1 is a member of the large family of ILT molecules (Immunologulin-like Transcripts) also known as Leukocyte Ig-like Receptors (LIRs) and Monocyte/Macrophage Ig-like Receptors (MIRs). ILT1 contains a short cytoplasmic domain. Through the cytoplasmic domain ILT1 associates with the Fc receptor gamma chain but not DAP12 resulting in a stimulatory signal. ILT1 may act as a receptor for MHC Class I antigens. Expression of ILT1 is mainly found on monocytes, macrophages, DC and weakly on granulocytes. ILT1 is thought to play a role in the TH1/Th2 balance as well as TLR signaling. The monoclonal antibody 135.4 has been reported to have activating properties upon crosslinking which result in calcium influx. Applications Reported: This 135.4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 135.4 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Leukocyte Ig-like receptors (LIRs) are a family of immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. All LIRs in subfamily B have an inhibitory function (LILRB1; MIM 604811). LIRs in subfamily A, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with trans membrane regions containing a charged arginine residue, may initiate stimulatory cascades. One member of subfamily A (LILRA3; MIM 604818) lacks a transmembrane region and is presumed to be a soluble receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD85h (ILT1)
Applications Flow Cytometry
Classification Monoclonal
Clone 135.4
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene LILRA2
Gene Accession No. Q8N149
Gene Alias CD85 antigen-like family member H; CD85h; ILT1; ILT-1; Immunoglobulin-like transcript 1; leucocyte Ig-like receptor A2; leukocyte immunoglobulin like receptor A2; leukocyte immunoglobulin-like receptor 7; Leukocyte immunoglobulin-like receptor subfamily A member 2; leukocyte immunoglobulin-like receptor subfamily A member 2 soluble; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 2; LILRA2; LIR7; LIR-7
Gene Symbols LILRA2
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11027
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.