Learn More
IBA Lifesciences CD81 Fab-Strep
Low affinity Fab fragment fused to a Twin-Strep-tag™ and specific for human CD81
Brand:  IBA Lifesciences 6-8015-150
Additional Details : Weight : 0.00600kg
Product Code. 17277600
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • CD81 Fab-Strep is a recombinantly expressed low affinity Fab fragment fused to a Twin-Strep-tag™
  • Due to its low affinity for the target, an increase in avidity is required to ensure sufficient binding to the cell or exosome surface
  • Combining Fab-Streps with a Strep-Tactin™ backbone leads to the formation of complexes that stably bind to cells or alternatively, also to exosomes
  • Fab-Streps were developed for Traceless Affinity Cell Selection (Fab-TACS™), which is based on our Strep-tag™ technology
Specifications
Specifications
Lyophilized
Fluorescent Cell Staining & Sorting, Magnetic or Affinity Chromatographic Cell Isolation
Human
Special Offers
For Research Use Only.