missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse anti-Human , APC-H7, Clone: HIT8a, BD Biosciences™
Mouse Monoclonal Antibody
128.50 EUR - 343.71 EUR
Specifications
| Antigen | CD8 |
|---|---|
| Clone | HIT8a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC-H7 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17287654
|
BD Biosciences
566856 |
25 Tests |
129.00 EUR 128.50 EUR / 25 tests Save 0.50 EUR 0.39% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17277654
|
BD Biosciences
566855 |
100 Tests |
344.00 EUR 343.71 EUR / 100 tests Save 0.29 EUR 0.08% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow CytometrySpecifications
| CD8 | |
| Flow Cytometry | |
| APC-H7 | |
| RUO | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with APC-H7 under optimum conditions, and unconjugated antibody and APC-H7 were removed. |
| HIT8a | |
| Monoclonal | |
| Mouse | |
| Human | |
| CD8α; CD8A; CD8 alpha; Leu2a; MAL; T8; p32 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title