Learn More
CD8 beta Antibody (CBB/1595), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-08335PE
Product Code. 18666608
  • 690.00 EUR / 0.10mL
Description
CD8 beta Monoclonal antibody specifically detects CD8 beta in Bovine, Goat, Sheep samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD8 beta
Monoclonal
PE
CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain
Antibody was raised against bovine CD8b. (Uniprot: P10966)
0.1 mL
Adaptive Immunity, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers
926
Store at 4°C in the dark.
IgG1 κ
ELISA
CBB/1595
PBS
Mouse
Protein A or G purified
RUO
Primary
Bovine, Goat, Sheep
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions