Learn More
Invitrogen™ CD68 Monoclonal Antibody (FA-11), PE
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  Invitrogen™ MA516678
Product Code. 15275939
  • 724.00 EUR / 100 tests
Description
Membrane permeabilization is required for flow cytometry applications. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. Rat anti Mouse CD68 antibody, clone FA-11 recognizes mouse macrosialin, a heavily glycosylated transmembrane protein and murine homolog of human CD68, which is classified as a unique scavenger receptor (ScR) family member, due to the presence of a lysosome associated membrane protein (LAMP)-like domain.
CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predominantly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human malignancies, including several Acute Myeloid Leukemia studies.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD68
Monoclonal
Conc. Not Determined
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
P31996
Cd68
purified Con A acceptor glycoprotein from P815 cell line.
100 Tests
Primary
Mouse
Antibody
IgG2a
Flow Cytometry
FA-11
PE
Cd68
CD68; CD68 antigen; CD68 molecule; GP110; Lamp4; macrophage antigen CD68; macrosialin; Scard1; scavenger receptor class D, member 1; similar to CD68 antigen
Rat
Protein G
RUO
12514
4°C, store in dark, DO NOT FREEZE!
Lyophilized