missing translation for 'onlineSavingsMsg'
Learn More

CD62L (L-selectin) Rat, anti-Mouse, PE, Clone: MEL-14, STEMCELL Technologies

Rat Monoclonal Antibody

POA - 321.30 EUR

Specifications

Antigen CD62L (L-selectin)
Clone MEL-14
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay
Classification Monoclonal
Conjugate PE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17169081
Encompass_Preferred
View Documents
Stemcell Technologies
60109PE.1
25 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17159081
Encompass_Preferred
View Documents
Stemcell Technologies
60109PE
100 μg
321.30 EUR
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The MEL-14 antibody reacts with murine CD62L (L-Selectin or LECAM-1), an ∼74 - 95 kDa type I glycoprotein expressed on the surface of a majority of leukocytes
  • CD62L mediates leukocyte-endothelial cell interactions through its association with ligands such as CD34, glyCAM-1, MAdCAM-1 and PSGL-1
  • It functions to facilitate lymphocyte rolling on the vascular endothelium, trafficking of lymphocytes to the lymph nodes, and homing of lymphocytes and neutrophils to sites of inflammation
  • Murine CD62L is expressed on most thymocytes, the highest expression levels being found on subsets of immunocompetent or dividing progenitor cells, as well as on neutrophils, eosinophils, monocytes, and subsets of B, T and NK cells
  • The MEL-14 antibody may be used together with antibodies to other cell surface markers to distinguish naive, memory and effector T cells
  • The MEL-14 antibody has been shown to block migration of lymphocytes to lymph nodes and to inhibit leukocyte rolling
Specifications

Specifications

CD62L (L-selectin)
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay
PE
Mouse
Primary
MEL-14
Monoclonal
Rat
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.