Learn More
CD3e (T-Cell Marker) Mouse, Clone: C3e/1931, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 916-MSM11-P1
This item is not returnable.
View return policy
Specifications
| CD3e (T-Cell Marker) | |
| Cell Surface and Cytoplasmic | |
| C3e/1931 | |
| Unconjugated | |
| Mouse | |
| 20kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 916 | |
| IgG1 κ |
| Immunohistochemistry, ELISA | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human CD3e fragment (apprx. aa 23-119) | |
| Jurkat cells. Tonsil or lymph node | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |