missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Rat anti-Mouse, BV711, Clone: RAM34, BD Biosciences™
Description
CD34 Monoclonal antibody specifically detects CD34 in Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | CD34 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | RAM34 |
| Conjugate | Brilliant Violet 711 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Cd34; CD34 antigen; Hematopoietic progenitor cell antigen CD34 |
| Host Species | Rat |
| Immunogen | Recombinant Mouse CD34 |
| Quantity | 50 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?