missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD33 Monoclonal Antibody (9A11-CD33), Super Bright™ 436, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 62-0331-80
This item is not returnable.
View return policy
Description
Rat Monoclonal Antibody
CD33 Monoclonal antibody specifically detects CD33 in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD33 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Q63994 | |
| CD33 | |
| Affinity chromatography | |
| RUO | |
| 12489 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 9A11-CD33 | |
| Super Bright 436 | |
| CD33 | |
| Cd33; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; Sialic acid-binding Ig-like lectin 3; SIGLEC3; SIGLEC-3 | |
| Rat | |
| 25 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction