missing translation for 'onlineSavingsMsg'
Learn More

CD31 Mouse anti-Human, Brilliant Violet 421, Clone: MBC 78.2 (also known as PECAM-1.2; 1.2), BD Optibuild™

Product Code. 15818759
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15818759 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15818759 Supplier BD Biosciences Supplier No. 744074

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The MBC 78.2 monoclonal antibody recognizes CD31 which is also known as, Platelet endothelial cell adhesion molecule (PECAM-1), platelet GPIIa, or EndoCAM. CD31 is a ñ130 kDa type I transmembrane glycoprotein that belongs to the Ig gene superfamily. CD31 is comprised of an extracellular region with six IgC-like domains, a transmembrane region, and a cytoplasmic domain that contains two immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The MBC 78.2 antibody specifically binds to an epitope located on membrane-proximal, extracellular Ig-like domain 6 of CD31. This epitope remains expressed by activated T cells after enzymatic cleavage and shedding of a soluble extracellular CD31 fragment comprised of Ig-like domains 1 to 5 from cells. In contrast to the MBC 78.2 antibody, the WM59 monoclonal antibody reportedly binds to the extracellular Ig-like domain 2 of CD31. WM59 can thus bind to cells that express intact CD31 but not to cells that express a truncated form CD31 that lacks at least the membrane distal Ig-like domains 1 and 2 of CD31. CD31 has wide tissue distribution and is expressed on platelets, monocytes, granulocytes, some T cell subsets, and at high levels on endothelial cells. This cell adhesion molecule has been implicated in a number of cellular phenomena, including vascular wound healing. angiogenesis, transendothelial migration of leucocytes, and the regulation of T cell responses.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD31
Classification Monoclonal
Clone MBC 78.2 (also known as PECAM-1.2; 1.2)
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias PECAM-1, PECAM1, EndoCAM, GPIIA'
Host Species Mouse
Immunogen Human CD31
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.