missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD307a (FCRL1) Mouse anti-Human, BUV737, Clone: E3, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 748655
This item is not returnable.
View return policy
Description
CD307a (FCRL1) Monoclonal antibody specifically detects CD307a (FCRL1) in Human samples. It is validated for Flow Cytometry
Specifications
| CD307a (FCRL1) | |
| Monoclonal | |
| Brilliant Ultraviolet 737 | |
| FCRL1; FCRH1; IFGP1; IRTA5; Immunoglobulin superfamily Fc receptor gp42 | |
| Human FCRL1 cDNA | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| E3 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu