missing translation for 'onlineSavingsMsg'
Learn More

CD300LG Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16854594 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16854594 20 μL 20µL
16844594 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16854594 Supplier Proteintech Supplier No. 154261AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CD300g receptor which may mediate L-selectin-dependent lymphocyte rollings. Binds SELL in a calcium dependent manner. Binds lymphocyte.

Specifications

Antigen CD300LG
Applications Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CD300LG
Gene Accession No. Q6UXG3
Gene Alias CD300LG, CLM 9, CLM9, CMRF35 like molecule 9, NEPMUCIN, TREM 4, TREM4
Gene Symbols CD300LG
Host Species Rabbit
Immunogen CD300LG Fusion Protein Ag8075
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146894
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.