missing translation for 'onlineSavingsMsg'
Learn More

CD3/CD20 Antibody Cocktail, Invitrogen™

Product Code. 11315063
Change view
Click to view available options
Quantity:
7 mL
Unit Size:
7mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11315063 7 mL 7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11315063 Supplier Invitrogen Supplier No. MA514433

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Cocktail

MA5-14433 targets CD3/CD20 in IHC (P) and WB applications and shows reactivity with Human, Mouse and Rat samples. CD3 (clone SP7) is a rabbit monoclonal IgG antibody. CD20 (clone L26) is an IgG2a mouse monoclonal antibody. The MA5-14433 immunogen is synthetic 13-mer peptide corresponding to aa 156-168 of the epsilon chain of human CD3 protein; the CD20 immunogen is human tonsil B cells.

The CD3 antibody reacts with the intracytoplasmic portion of the CD3 antigen expressed by normal and neoplastic T-lymphocytes. It stains human T-lymphocytes in both cortex and medulla of the thymus and in peripheral lymphoid tissues. CD20 is a non-Ig differentiation antigen of B-lymphocytes and the expression of CD20 is restricted to normal and neoplastic B-lymphocytes, being absent from all other leukocytes and tissues. The CD3-CD20 cocktail reveals lymphoid architecture in neoplastic situations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3/CD20
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Cocktail
Clone CD3-SP7; CD20-L26
Conjugate Unconjugated
Formulation tris HCl with proprietary stabilizer and <0.1% sodium azide; pH 7.6
Gene MS4A1
Gene Accession No. P04234, P04235, P07766, P09693, P11836, P11942, P19377, P19437, P22646, Q64159
Gene Alias AA960661; AI504783; B1; B-cell differentiation antigen Ly-44; B-lymphocyte antigen CD20; B-lymphocyte cell-surface antigen B1; B-lymphocyte surface antigen B1; Bp35; CD20; CD20 antigen; CD20 receptor; CD3; CD3 antigen delta polypeptide; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 antigen, epsilon polypeptide; CD3 antigen, gamma polypeptide; CD3 delta; CD3 gamma-chain; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3 molecule, epsilon; CD3 molecule, epsilon polypeptide; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; Cd3d; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule; CD3d molecule, delta (CD3-TCR complex); CD3-DELTA; Cd3e; CD3e antigen, epsilon polypeptide (TiT3 complex); CD3e molecule; CD3e molecule, epsilon (CD3-TCR complex); CD3epsilon; CD3-epsilon; CD3g; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule; CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3-GAMMA; Ctg3; Ctg-3; CVID5; IMD17; IMD18; IMD19; LEU-16; Leukocyte surface antigen Leu-16; Ly-44; Lymphocyte antigen 44; membrane spanning 4-domains A1; membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 1; membrane-spanning 4-domains, subfamily A, member 2; Ms4a1; Ms4a2; OKT3, delta chain; S7; T3d; T3e; T3G; T-cell antigen receptor complex, epsilon subunit of T3; T-cell antigen receptor complex, gamma subunit of T3; T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 delta chain; T-cell surface glycoprotein CD3 epsilon chain; T-cell surface glycoprotein CD3 gamma chain; TCRE
Gene Symbols CD3D, Cd3e, CD3G, MS4A1
Host Species Mixed, Rabbit
Immunogen synthetic 13-mer peptide corresponding to aa 156-168 of the epsilon chain of human CD3 protien; CD20- Human tonsil B cells
Purification Method purified
Quantity 7 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12482, 12500, 12501, 12502, 25710, 300678, 309217, 315609, 915, 916, 917, 931
Target Species Human, Mouse, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.