missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD249 (Ly-51, 6C3/BP-1 antigen) Mouse anti-Mouse, BUV615, Clone: BP-1, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 752355
This item is not returnable.
View return policy
Description
CD249 (Ly-51, 6C3/BP-1 antigen) Monoclonal antibody specifically detects CD249 (Ly-51, 6C3/BP-1 antigen) in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD249 (Ly-51, 6C3/BP-1 antigen) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Ms CD249 (Ly-51) BUV615 BP-1 50 μg | |
| Abelson murine leukemia virus-transformed BALB/c mouse pre-B cell line 18.81 | |
| RUO | |
| CD249 (Ly-51, 6C3/BP-1 antigen) | |
| Refrigerate (2 to 8°C) | |
| Mouse |
| Flow Cytometry | |
| BP-1 | |
| Brilliant Ultraviolet 615 | |
| Mouse | |
| 1 x 50 μg | |
| Primary | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction