missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The T cell antigen receptor (TCR) consists of a ligand-specific alpha/beta heterodimer non-covalently associated with five invariant chains including the CD3 gamma/delta/eta and zeta subunits, all of which are required for efficient surface expression. T cell activation through the TCR induces cellular differentiation and/or proliferation and the production of lymphokines and cytokines. Both the CD3 and TCR zeta subunits are proposed to be responsible for the intracellular signal transduction events.
Specifications
Specifications
| Antigen | CD247 |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | CD247 |
| Gene Accession No. | P20963, P24161 |
| Gene Alias | CD247, CD247 molecule, CD3 ZETA, CD3H, CD3Q, CD3Z, T cell receptor T3 zeta chain, T3Z, TCRZ |
| Gene Symbols | CD247 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?