Learn More
CD1d Mouse, anti-Human, CoraLite Plus 488, Clone: 1C12F1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66257-100UL
 View more versions of this product
Product Code. 18025145
  • 450.00 EUR / 100µL
Description
The CD1d antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD1d. This antibody recognizes human antigen. The CD1d antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
CD1d
1C12F1
CoraLite Plus 488
Mouse
Primary
Human
155 kDa
IgG1
Western Blot
1000 μg/mL
CD1A, CD1D, CD1d molecule, R3, R3G1
100 μL
912
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.