missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Antibody (FMC63) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
284.00 EUR - 558.00 EUR
Specifications
| Antigen | CD19 |
|---|---|
| Clone | FMC63 |
| Dilution | Flow Cytometry 1:10 - 1:1000, Immunofluorescence 1:10 - 1:500, CyTOF-ready |
| Applications | Flow Cytometry, Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18220011
|
Novus Biologicals
NBP2-52688-0.025MG |
0.025 mg |
284.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18242270
|
Novus Biologicals
NBP2-52688-0.2MG |
0.2 mg |
558.00 EUR
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, CyTOF-ready.Specifications
| CD19 | |
| Flow Cytometry 1:10 - 1:1000, Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
| CD19 | |
| IgG | |
| Protein A or G purified | |
| Binds human CD19. |
| FMC63 | |
| Flow Cytometry, Immunofluorescence, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| 930 | |
| Human prolymphocytic leukaemia cell line JVM3. | |
| Primary | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title