Learn More
CD161f Rat anti-Mouse, Brilliant Violet 421, Clone: C8, BD
Product Code. 15844929
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15844929 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15844929 Supplier BD Biosciences Supplier No. 566304
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Rat Monoclonal Antibody
The C8 monoclonal antibody specifically recognizes C57BL/6 mouse CD161 antigen-like family member F (CD161f ). CD161f is also known as Natural killer cell surface protein NKR-P1F (Nkrp1f), and is encoded by Klrb1f /(Killer cell lectin-like receptor subfamily B member 1F). CD161f is expressed on natural killer (NK) cells, and some dendritic cells and activated lymphocytes. CD161f binds to C-type lectin related protein c, d, and g (Clr-c, d, g) which are expressed on several cell types including dendritic cells, B cells, and activated T cells. CD161f-mediated signaling can reportedly costimulate T cell proliferation and IL-2 production, and may play a role in dendritic cell maturation.
The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY
Specifications
Antigen CD161f
Classification Monoclonal
Clone C8
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Klrb1f, Nkrp1-f, Nkrp1f, NKR-P1F
Host Species Rat
Immunogen Mouse NKRP1F Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ