missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Antibody (M5E2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 11 publications
Brand: Novus Biologicals NB100-77758
272.85 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
CD14 Monoclonal specifically detects CD14 in Human, Porcine, Bovine, Canine, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, CyTOF-ready, Block/Neutralize, Dual RNAscope ISH-IHC.
Specifications
| CD14 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Simple Western 50 ug/ml, Flow Cytometry < 2.0 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-ready, Block/Neutralize reported in scientific literature (PMID 27228163), Dual RNAscope ISH-IHC | |
| CD14 | |
| Human peripheral blood cells. | |
| 0.1 mg | |
| Apoptosis, Asthma, Cancer, Cell Biology, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers | |
| 929 | |
| Human, Pig, Bovine, Canine, Primate | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| M5E2 | |
| Unconjugated | |
| CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| CD14 (M5E2). Expected to cross react with Chimpanzee, Cynomolgus, Rhesus, Pigtailed Macaque, Capuchin Monkey, Cotton-topped Tamarin, Common Marmoset, Squirrel Monkey. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction