Learn More
CD137 Ligand (4-1BB Ligand) Monoclonal Antibody (5G11), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 14-9056-80
Description
This 5G11 monoclonal antibody recognizes human CD137 ligand also known as 4-1BB ligand, TNFSF9, or CDw137L. CD137 ligand is a member of the TNF superfamily, and is a type II membrane protein. It is a co-stimulatory molecule that is expressed on antigen presenting cells (DCs, monocytes/macrophages, B cells) and is upregulated upon activation of these cells. Its receptor, CD137 (4-1BB) is found on a variety of cells, including inflamed endothelial cells, where it enhances extravasation of CD137 ligand-expressing monocytes. Activated T cells also express CD137, and engagement with CD137 ligand enhances T cell proliferation, IL-2 secretion, survival, and cytotoxic activity. CD137 ligand is also expressed on a majority of B-cell lymphomas, including mantle cell lymphoma, follicular lymphoma, and diffuse large B-cell lymphoma (but not on Hodgkin lymphoma and T-cell lymphoma). This 5G11 antibody has been tested by immunohistochemical staining of formalin-fixed paraffin embedded human tissue using low pH antigen retreival and can be used at less than or equal to 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This transmembrane cytokine is a bidirectional signal transducer that acts as a ligand for TNFRSF9/4-1BB, which is a costimulatory receptor molecule in T lymphocytes. This cytokine and its receptor are involved in the antigen presentation process and in the generation of cytotoxic T cells. The receptor TNFRSF9/4-1BB is absent from resting T lymphocytes but rapidly expressed upon antigenic stimulation. The ligand encoded by this gene, TNFSF9/4-1BBL, has been shown to reactivate anergic T lymphocytes in addition to promoting T lymphocyte proliferation. This cytokine has also been shown to be required for the optimal CD8 responses in CD8 T cells. This cytokine is expressed in carcinoma cell lines, and is thought to be involved in T cell-tumor cell interaction.
Specifications
| CD137 Ligand (4-1BB Ligand) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| P41273 | |
| TNFSF9 | |
| Affinity chromatography | |
| RUO | |
| 8744 | |
| 4° C, do not freeze | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 5G11 | |
| Unconjugated | |
| TNFSF9 | |
| TNFSF9, CDw137L, CD137L, 4-1BBL, 41BBL | |
| Mouse | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only