missing translation for 'onlineSavingsMsg'
Learn More

CD108 Mouse anti-Human, BUV805, Clone: KS-2, BD Biosciences™

Mouse Monoclonal Antibody

Brand:  BD Biosciences 749451

Product Code. 17201005

  • ONLINE EXCLUSIVE PRICE
    540.00 EUR
    539.91 EUR / 50µg
    Save 0.09 EUR
    0.02% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD108 Monoclonal antibody specifically detects CD108 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD108
Monoclonal
Brilliant Ultraviolet 805
JMH; SEM7A; Sema K1; Sema L; John-Milton-Hargen human blood group Ag
BALB/c splenocytes immunized with PHA-activated human PBMCs fused with murine cell line NS-1
RUO
Human
IgG2a κ
Flow Cytometry
KS-2
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.