missing translation for 'onlineSavingsMsg'
Learn More

CCT251236, MedChemExpress

Product Code. 16470719
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16470719

Brand: Medchem Express HY1010261mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CCT251236 is an orally available pirin ligand from a heat shock transcription factor 1 (hsf1) phenotypic screen with an IC50 of 19 nM for inhibition of HSF1-mediated HSP72 induction.

CCT251236 is an orally available pirin ligand from a heat shock transcription factor 1 (hsf1) phenotypic screen with an IC50 of 19 nM for inhibition of HSF1-mediated HSP72 induction.

Specifications

Chemical Name or Material CCT251236
CAS 1693731-40-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C32H32N4O5
Quantity 1 mg
Solubility Information DMSO : ≥ 150 mg/mL (271.43 mM)
SMILES CC1=CC=C(NC(C2=CC(OCCO3)=C3C=C2)=O)C=C1NC(C4=CC(C=CC(OCCN5CCCC5)=N6)=C6C=C4)=O
Molecular Weight (g/mol) 552.62
Formula Weight 552.62
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.