Learn More
CCT020312, MedChemExpress
Product Code. 16467647
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467647 5 mg 5mg
16441147 50 mg 50mg
16425676 100 mg 100mg
16414898 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16467647 Supplier Medchem Express Supplier No. HY119240
MedChemExpress CCT020312 is a selective EIF2AK3/PERK activator. CCT020312 elicits EIF2A phosphorylation in cells.
CCT020312 is a selective EIF2AK3/PERK activator. CCT020312 elicits EIF2A phosphorylation in cells.
Specifications
Chemical Name or Material CCT020312
CAS 324759-76-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H30Br2N4O2
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (153.75 mM; Need ultrasonic)
SMILES O=C1NC2=C(C(C3=CC=CC=C3)=C1C4=NN(C(C4)C5=CC=C(C=C5)Br)C(CCN(CC)CC)=O)C=C(C=C2)Br
Molecular Weight (g/mol) 650.4
Formula Weight 650.4
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease