Learn More
Paragon Scientific™ CCS Viscosity Standards
Product Code. 15508864
Click to view available options
Type:
SAE Specification J303
SAE Specification J306
SAE Specification J308
SAE Specification J309
SAE Specification J310
SAE Specification J311
SAE Specification J312
SAE Specification J314
SAE Specification J315
SAE Specification J316
SAE Specification J317
SAE Specification J318
SAE Specification J319
SAE Specification J320
SAE Specification J321
SAE Specification J322
SAE Specification J323
SAE Specification J324
SAE Specification J325
SAE Specification J326
SAE Specification J328
Unit Size:
500mL
Product Code. 15508864
Brand: Paragon Scientific™ CL160
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Paragon ISO 17025 / ISO 17034 certified CCS viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing to ASTM D5293 and SAE Specification J300.
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Full Traceability to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Manufactured in the United Kingdom
  • 2 year shelf life
Specifications
Certifications/Compliance Full Traceability to National Standards
Type SAE Specification J310
Quantity 500 mL
Nominal values quoted