missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL3/MIP-1 alpha Rat anti-Mouse, Clone: 39624, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB450-100
This item is not returnable.
View return policy
Description
CCL3/MIP-1 alpha Monoclonal specifically detects CCL3/MIP-1 alpha in Mouse samples. It is validated for Western Blot.Specifications
| CCL3/MIP-1 alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6348 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 39624 | |
| Western Blot 1 ug/mL | |
| P10855 | |
| CCL3 | |
| E. coli-derived recombinant mouse CCL3/MIP-1 alpha Ala24-Ala92 Accession # P10855 | |
| 100 μg | |
| Primary | |
| Detects mouse CCL3/MIP-1 alpha in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant mouse CCL4/ MIP-1 beta is observed and no cross-reactivity with recombinant human CCL1, 2, 3, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 21, 22, 23, 24, 25, 28, recombinant mouse CCL1, 2, 6, 7, 9/10/MIP-1 gamma , 11, 12, 19, 21, 22, 25, 28, recombinant rat CCL2 or CCL20 is observed. | |
| Mouse | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?