missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL11/Eotaxin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
287.00 EUR - 840.00 EUR
Specifications
| Antigen | CCL11/Eotaxin |
|---|---|
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18669808
|
Novus Biologicals
NBP3-11356-0.025mg |
0.025 mg |
287.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692999
|
Novus Biologicals
NBP3-11356-0.1mg |
0.1 mg |
840.00 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL11/Eotaxin Polyclonal antibody specifically detects CCL11/Eotaxin in Porcine samples. It is validated for Western Blot, ELISASpecifications
| CCL11/Eotaxin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Pig | |
| C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin) | |
| Recombinant porcine Eotaxin 1 | |
| Primary | |
| Store at 4°C. Do not freeze. |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Asthma, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology | |
| PBS | |
| 6356 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title