Learn More
CBS Antibody (5F7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00000875-M07
This item is not returnable.
View return policy
Description
CBS Monoclonal antibody specifically detects CBS in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CBS | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| cystathionine-beta-synthase, EC 4.2.1.2210Beta-thionase | |
| CBS (NP_000062, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPSETPQAEVGPTGCPHRSGPHSAKGSLEKGSPEDKEAKEPLWIRPDAPSRCTWQLGRPASESPHHHTAPAKSPKILPDILKKIGDTPMVRINKIGKKFG | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 875 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 5F7 | |
| Western Blot 1:500 | |
| NP_000062 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |