missing translation for 'onlineSavingsMsg'
Learn More

Caveolin-3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Product Code. 16866105 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16866105 20 μL 20µL
16856105 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16866105 Supplier Proteintech Supplier No. 283581AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The Caveolin-3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Caveolin-3. This antibody recognizes human, mouse, rat antigen. The Caveolin-3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Antigen Caveolin-3
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 467 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 859
Target Species Human, Rat, Mouse
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.