Learn More
Brady™ Caution Labels
Product Code. 11726835
Click to view available options
Message:
DANGER DEFENSE D’ENCLENCHER TANT QUE L’ETIQUETTE EST APPLIQUEE
Length (Metric):
50 mm
Width (Metric):
50 mm
Unit Size:
1 pack
Product Code. 11726835
Brand: Brady™ 225256
Only null left
Specifications
Quantity 10 Pk.
Type Tag
Alternate Part Number TAG-DEFENSE DENCLEN.-50X110MM
Color Black on Yellow
Material Polyester
Message DANGER DEFENSE D’ENCLENCHER TANT QUE L’ETIQUETTE EST APPLIQUEE
For Use With (Application) Lockout Tagout
Height (English) 4.33 in.
Height (Metric) 110 mm
Length (Metric) 50 mm
Length (English) 2 in.
Thickness 0.249 mm
Temperature Range (Metric) -40°C to +100°C
Temperature Range (English) -40°F to +212°F
Width (English) 1.97 in.
Width (Metric) 50 mm
Diameter (Metric) Hole 7 mm
Label Type Lockout Tagout Tags