Learn More
Thermo Scientific™ Barnstead™ Water System Cartridges
Product Code. p-4589241 Shop All Thermo Scientific Products
Click to view available options
Description:
High purity water cartridge
Ion exchange cartridge
Ultrapure polisher cartridge
For Use With (Equipment):
GenPure GenPure Pro, GenPure xCAD, LabTower EDI
MicroPure
Pacific TII
Smart2Pure
Unit Size:
Each
Product Code. 11397151
Brand: Thermo Scientific™ 09.4011
Maintain your Thermo Scientific™ Barnstead™ point of use water purification system with these replacement cartridges.
Maintain your Thermo Scientific™ Barnstead™ point of use water purification systems with replacement Thermo Scientific™ Barnstead™ Cartridges.
Choose the appropriate part number for your system. These cartridges are for use with the following Barnstead point of use water purification systems:
  • MicroPure™
  • GenPure™
  • GenPure™ Pro
  • GenPure™ xCAD
  • Smart2Pure™
  • LabTower™ EDI
  • Pacific™ TII
TRUSTED_SUSTAINABILITY
Specifications
Description High purity water cartridge
For Use With (Equipment) Pacific TII
Type Cartridge, High-purity Water
Product Content Correction
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.