Learn More
CleanSpace™ Technology Carry Backpack
Product Code. 15900082 Shop All CleanSpace Technology Products
Click to view available options
:
Color: Black
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15900082
Brand: CleanSpace™ Technology PAF0099
Only null left
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Carry backpack for use with CleanSpace™ respirator systems.
  • Black
  • For use with all CleanSpace™ Respirators
Specifications
Color Black
Disposable Reusable
Product Type Backpack
For Use With (Equipment) Respirators
Model PAF-0099