Learn More
Motic™ Achromatic Oil Immersion Objective EA
Description
- Economy/Education Series Achromatic Objective
- Magnification: 100X
- WD: 0.85mm
- Oil immersion
- Spring loaded
Specifications
Specifications
| Type | Oil Immersion Objective |
| Focus Distance | 1.25 mm |
| For Use With (Equipment) | Motic™ 1802LED, 1820LED, 2801LED, 2802LED, 2820 LED, 2823LED model microscopes |
| Magnification Power | 100X |
| Quantity | 1 Piece |