missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Color | White |
| Quantity | 1 kg |
| IUPAC Name | 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium |
| Formula Weight | ≈90 kd |
| Physical Form | Powder |
Safety and Handling
Product Identifier
- Carboxymethyl Cellulose Sodium Salt, Low Viscosity
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Skin sensitiser Category 1
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Carboxymethyl Cellulose Sodium Salt
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?