missing translation for 'onlineSavingsMsg'
Learn More

Carboxy pyridostatin trifluoroacetate salt, MedChemExpress

Product Code. 16487387
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487387 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16487387 Supplier Medchem Express Supplier No. HY112680A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Carboxy pyridostatin trifluoroacetate salt has the peculiarity to exhibit high molecular specificity for RNA over DNA G4s.

Carboxy pyridostatin trifluoroacetate salt has the peculiarity to exhibit high molecular specificity for RNA over DNA G4s.

Specifications

Chemical Name or Material Carboxy pyridostatin trifluoroacetate salt
CAS 2444713-88-4
Color Light Green
Purity Grade Notes Research
Molecular Formula C37H35F3N10O9
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (121.84 mM; Need ultrasonic) ∣H2O : 60 mg/mL (73.11 mM; Need ultrasonic)
SMILES O=C(O)CCN1N=NC(COC2=CC(C(NC3=NC4=CC=CC=C4C(OCCN)=C3)=O)=NC(C(NC5=NC6=CC=CC=C6C(OCCN)=C5)=O)=C2)=C1.FC(C(O)=O)(F)F
Molecular Weight (g/mol) 820.73
Formula Weight 820.73
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.