Learn More
Invitrogen™ Carbonic Anhydrase XIII Polyclonal Antibody
GREENER_CHOICE
Product Code. 17741674
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17741674 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17741674 Supplier Invitrogen™ Supplier No. PA5120274
Rabbit Polyclonal Antibody
Positive test controls include: HT-29, DU145, Mouse spleen, Mouse lung, Mouse kidney, Mouse heart. Immunogen sequence: MSRLSWGYRE HNGPIHWKEF FPIADGDQQS PIEIKTKEVK YDSSLRPLSI KYDPSSAKII SNSGHSFNVD FDDTENKSVL RGGPLTGSYR LRQVHLHWGS ADDHGSEHIV DGVSYAAELH VVHWNSDKYP SFVEAAHEPD GLAVLGVFLQ IGEPNSQLQK ITDTLDSIKE KGKQTRFTNF DLLSLLPPSW DYWTYPGSLT VPPLLESVTW IVLKQPINIS.
Carbonic anhydrase XII (CA XII), encoded by the CA12 gene, is one of the enzyme isoforms in the carbonic anhydrase family that catalyzes the reversible hydration of carbon dioxide into bicarbonate and protons. CA XII is a transmembrane protein expressed in various tissues, including the kidneys, intestines, and lungs, playing crucial roles in maintaining acid-base balance, pH regulation, and electrolyte transport. Its activity is significant in organs where fluid secretion and absorption are essential processes, impacting functions such as respiration and renal bicarbonate reabsorption. CA XII is involved in physiological processes like cell proliferation and differentiation by modulating the extracellular and intracellular pH, influencing tumor microenvironment dynamics. Aberrant expression of CA XII has been noted in several cancers, including breast and colorectal cancers, where it supports tumor growth and metastasis by maintaining the acid-base homeostasis conducive to cancer cell survival. Due to its surface expression and role in maintaining pH balance, CA XII is a target of interest for therapeutic interventions aiming to disrupt tumor progression and enhance the efficacy of cancer treatments.
TRUSTED_SUSTAINABILITY
Specifications
Antigen Carbonic Anhydrase XIII
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ca13
Gene Accession No. Q8N1Q1, Q9D6N1
Gene Alias 2310075C21Rik; Ca13; Car13; Carbonate dehydratase XIII; carbonic anhydrase 13; Carbonic anhydrase XIII; CAXIII; CA-XIII; RGD1560453
Gene Symbols Ca13, Car13
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human CA13 (NP_940986.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 377677, 71934
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG