missing translation for 'onlineSavingsMsg'
Learn More

Carbamoyl-guanidine Amidino Urea, TRC

Product Code. 30004799
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004799 500 mg 500mg
30137773 10 g 10g
30010680 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004799 Supplier Toronto Research Chemicals Supplier No. TRCC175873500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 926-72-7
Molecular Formula C2 H6 N4 O . Cl H
Molecular Weight (g/mol) 138.56
Synonym Urea, N-(aminoiminomethyl)-, hydrochloride (1:1), Urea, (aminoiminomethyl)-, monohydrochloride (9CI), Urea, amidino-, hydrochloride (7CI), Urea, amidino-, monohydrochloride (8CI), Guanylurea hydrochloride
IUPAC Name carbamimidoylurea;hydrochloride
SMILES Cl.NC(=N)NC(=O)N

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C2H6N4O.ClH/c3-1(4)6-2(5)7;/h(H6,3,4,5,6,7);1H
Formula Weight 138.0308
Chemical Name or Material Carbamoyl-guanidine Amidino Urea Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.