missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPZB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25043-1-AP-20UL
This item is not returnable.
View return policy
Description
CAPZB is a member of the F-actin capping protein family. This gene encodes the beta subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.Specifications
| CAPZB | |
| Polyclonal | |
| Unconjugated | |
| CAPZB | |
| CAPB, CAPPB, CAPZ, CapZ beta, CAPZB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12345, 832 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.51 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P47756, P47757 | |
| CAPZB | |
| CAPZB Fusion Protein Ag19327 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction