missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CAPZA2 Polyclonal Antibody

Product Code. 15730391
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15730391 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15730391 Supplier Invitrogen™ Supplier No. PA552438

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: IEGYEDQVLI TEHGDLGNGK FLDPKNRICF KFDHLRKEAT DPRPCEVENA VESWRTSVET ALRAYVKEHY PNGVCTVYGK KIDGQQTIIA CIESHQFQA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

The protein encoded by this gene is a member of the F-actin capping protein alpha subunit family. It is the alpha subunit of the barbed-end actin binding protein Cap Z. By capping the barbed end of actin filaments, Cap Z regulates the growth of the actin filaments at the barbed end.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAPZA2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Capza2
Gene Accession No. P47755
Gene Alias 1110053K06Rik; AW208808; Cappa2; capping actin protein of muscle Z-line alpha subunit 2; capping protein (actin filament) muscle Z-line, alpha 2; capping protein alpha 2; CAPZ; capZ alpha-2; CAPZA2; CAZA2; F-actin capping protein alpha-2 subunit; F-actin-capping protein subunit alpha-2
Gene Symbols Capza2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CAPZA2. Recombinant protein control fragment (Product #RP-89942).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 830
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.